Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

737127

Sigma-Aldrich

(S)-(−)-2-Methyl-2-propanesulfinamide solution

1 M in THF, contains BHT as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NOS
Molecular Weight:
121.20
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

liquid

optical activity

[α]22/D -1.0°, c = 0.5 in chloroform

contains

BHT as inhibitor

concentration

1 M in THF

refractive index

n20/D 1.418

density

0.895 g/mL at 25 °C

SMILES string

CC(C)(C)S(N)=O

InChI

1S/C4H11NOS/c1-4(2,3)7(5)6/h5H2,1-3H3/t7-/m0/s1

InChI key

CESUXLKAADQNTB-ZETCQYMHSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-20.0 °F

flash_point_c

-28.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service