Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

736562

Sigma-Aldrich

Lithium-6Li bistrifluoromethanesulfonimidate

95 atom % 6Li, 98% (CP)

Synonym(s):

Bis(trifluoromethane)sulfonimide lithium-6Li salt, Bis(trifluoromethylsulfonyl)amine lithium-6Li salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO2N6LiSO2CF3
Molecular Weight:
286.16
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

95 atom % 6Li

assay

98% (CP)

mp

234-238 °C (lit.)

mass shift

M-1

SMILES string

[6Li]N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

InChI

1S/C2F6NO4S2.Li/c3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q-1;+1/i;1-1

InChI key

QSZMZKBZAYQGRS-HCMAANCNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service