736295
Poly(3,4-ethylenedioxythiophene), bis-poly(ethyleneglycol), lauryl terminated
0.7 wt. % (dispersion in nitromethane), contains p-toluenesulfonate as dopant
Synonym(s):
Aedotron™ P3-NM, C12-PEG-block-PEDOT-block-PEG-C12, PEDOT:PEG
About This Item
Recommended Products
form
liquid (dispersion)
contains
p-toluenesulfonate as dopant
composition
Aedotron™ TM-P3 polymer, 0.8-1.2 wt. %
ethanol, 4-8 wt. %
isopropanol, 0.2-0.8 wt. %
nitromethane, 90-95 wt. %
concentration
0.5-0.9 wt. % (solid concentration)
0.7 wt. % (dispersion in nitromethane)
refractive index
n20/D 1.381
conductivity
10−2-10−4 S/cm (bulk)
density
1.107 g/mL at 25 °C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
signalword
Warning
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
78.1 °F - closed cup
flash_point_c
25.6 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Progress in Organic Thermoelectric Materials & Devices including high ZT values of >0.2 at room temperature by p-type (PEDOT:PSS) & n-type (Poly[Kx(Ni-ett)]) materials are discussed.
The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.
Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service