Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

736058

Sigma-Aldrich

2-Fluoropyridine-3-carbonitrile

97%

Synonym(s):

3-Cyano-2-fluoropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3FN2
CAS Number:
Molecular Weight:
122.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

28-33 °C

SMILES string

Fc1ncccc1C#N

InChI

1S/C6H3FN2/c7-6-5(4-8)2-1-3-9-6/h1-3H

Inchi Key

USIDQCCXMGJOJM-UHFFFAOYSA-N

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

210.0 °F

flash_point_c

98.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service