Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735833

Sigma-Aldrich

(S)-1-Boc-3-(methylamino)piperidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O2
CAS Number:
Molecular Weight:
214.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.467

density

0.985 at 25 °C

storage temp.

2-8°C

SMILES string

CN[C@H]1CCCN(C1)C(=O)OC(C)(C)C

InChI

1S/C11H22N2O2/c1-11(2,3)15-10(14)13-7-5-6-9(8-13)12-4/h9,12H,5-8H2,1-4H3/t9-/m0/s1

InChI key

XRRRUOWSHGFPTI-VIFPVBQESA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service