Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735019

Sigma-Aldrich

1-(Phenylsulfonyl)-7-azaindole-3-boronic acid pinacol ester

95%

Synonym(s):

1-(Phenylsulfonyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- 1H-pyrrolo[2,3-b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21BN2O4S
CAS Number:
Molecular Weight:
384.26
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

355-360 °C

SMILES string

CC1(C)OB(OC1(C)C)c2cn(c3ncccc23)S(=O)(=O)c4ccccc4

InChI

1S/C19H21BN2O4S/c1-18(2)19(3,4)26-20(25-18)16-13-22(17-15(16)11-8-12-21-17)27(23,24)14-9-6-5-7-10-14/h5-13H,1-4H3

InChI key

KZANVIJXSQABKR-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service