Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

734675

Sigma-Aldrich

Cerium(IV) oxide-samarium doped

greener alternative

pellets (for sputtering), contains 15 mol % samarium as dopant

Synonym(s):

SDC-15, SDC, Samarium-doped ceria

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeO2
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

pellets (for sputtering)

contains

15 mol % samarium as dopant

composition

Sm0.15Ce0.85O2

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

diam. × L

2-3 mm × 3-5 mm

greener alternative category

SMILES string

[Sm].O=[Ce]=O

InChI

1S/Ce.2O.Sm

Inchi Key

AQGZISMLQKBRRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cerium(IV) oxide-samarium doped pellets (15% mol samarium doped) (SDC-15) is an electrolytic material with a high oxygen ion conductivity. Samarium ion acts as an aliovalent cation that can be incorporated into the host lattice to increase the oxygen vacancy concentration and improve the oxide ion conduction in cerium oxide. It can be synthesized by reverse strike co-precipitation technique.[1][2]
Samarium doped ceria may be used as an electrolyte in solid oxide fuel cells (SOFCs).[1][3] Electrical conductivity as a function of temperature of doped ceria was investigated in a study.[2] Relation of dopant type, oxygen vacancy ordering and ionic conductivity was investigated by transmission electron microscopy and electron energy loss spectroscopy.[4]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Cerium(IV) oxide-samarium doped can be used in the fabrication of solid oxide fuel cells (SOFCs), which can further be used as sustainable sources of alternative energy.[5][6]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intermediate-temperature electrochemical performance of a polycrystalline PrBaCo2O5+ δ cathode on samarium-doped ceria electrolyte.
Chen D, et al.
Journal of Power Sources, 188(1), 96-105 (2009)
Stephan Immenschuh et al.
Transplantation, 99(1), 56-62 (2014-08-15)
Treatment of patients with antibody-mediated rejection (AMR) after kidney transplantation by rituximab and plasmapheresis is ambiguous. Because of its unknown efficiency and serious side effects, biomarkers, which are predictive for responsiveness to this treatment in AMR patients, are required. Twenty
AC impedance investigation of samarium-doped ceria."
Zhan Z, et al.
Journal of the Electrochemical Society, 148(5), A427-A432 (2001)
Harsharan K Singh et al.
Transplantation, 99(3), 609-615 (2014-08-20)
A qualitative highly predictive urinary test for polyomavirus nephropathy (PVN) is the PV-Haufen test. This article evaluates whether a quantitative PV-Haufen analysis, that is, the number of PV-Haufen shed per milliliter urine, predicts PVN disease grades and the severity of
Diego H Sanchez et al.
PLoS genetics, 10(11), e1004806-e1004806 (2014-11-21)
Epigenetic mechanisms suppress the transcription of transposons and DNA repeats; however, this suppression can be transiently released under prolonged heat stress. Here we show that the Arabidopsis thaliana imprinted gene SDC, which is silent during vegetative growth due to DNA

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service