Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

734535

Sigma-Aldrich

(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

95%

Synonym(s):

Crabtree′s catalyst BARF analog

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H62BF24IrNP
Molecular Weight:
1523.14
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

reaction suitability

core: iridium
reagent type: catalyst

mp

168-173 °C

storage temp.

−20°C

SMILES string

C[Ir+]C.c1ccncc1.C2CC=CCCC=C2.C3CCC(CC3)P(C4CCCCC4)C5CCCCC5.FC(F)(F)c6cc(cc(c6)C(F)(F)F)[B-](c7cc(cc(c7)C(F)(F)F)C(F)(F)F)(c8cc(cc(c8)C(F)(F)F)C(F)(F)F)c9cc(cc(c9)C(F)(F)F)C(F)(F)F

InChI

1S/C32H12BF24.C18H33P.C8H12.C5H5N.2CH3.Ir/c34-25(35,36)13-1-14(26(37,38)39)6-21(5-13)33(22-7-15(27(40,41)42)2-16(8-22)28(43,44)45,23-9-17(29(46,47)48)3-18(10-23)30(49,50)51)24-11-19(31(52,53)54)4-20(12-24)32(55,56)57;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-4-6-8-7-5-3-1;1-2-4-6-5-3-1;;;/h1-12H;16-18H,1-15H2;1-2,7-8H,3-6H2;1-5H;2*1H3;/q-1;;;;;;+1/b;;2-1-,8-7-;;;;

InChI key

OVABWJKBCHJIIP-OHHPTVSFSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Crabtree Group developed the [(cod)IrLL']PF6 series of catalysts, where L is a P-donor such as PCy3 and L' and N-donor such as pyridine. These are very active in the hydrogenation of sterically hindered alkenes. The catalyst also shows directing effects as a result of binding of the catalyst to suitable functional groups on the substrate, followed by addition of H2 from the same side of the substrate that the functional group is located. Two versions of the catalyst are available from us one with PF6 and the other with BArF4 counteranion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service