Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

734357

Sigma-Aldrich

2,4-Dihydroxypyrimidine-2,4,5-13C3-5-carboxylic acid-13C monohydrate

99 atom % 13C, 97% (CP)

Synonym(s):

Uracil-2,4,5-13C3-5-carboxylic acid-13C monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4CH4N2O4 · xH2O
Molecular Weight:
160.07 (anhydrous basis)
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

assay

97% (CP)

form

solid

mp

283 °C (dec) (lit.)

mass shift

M+4

SMILES string

O.O[13C](=O)[13c]1cn[13c](O)n[13c]1O

InChI

1S/C5H4N2O4.H2O/c8-3-2(4(9)10)1-6-5(11)7-3;/h1H,(H,9,10)(H2,6,7,8,11);1H2/i2+1,3+1,4+1,5+1;

InChI key

HUMYNECKRWOZGW-QWJITLNOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service