Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

734136

Sigma-Aldrich

Di-tert-butylferrocenylphosphonium tetrafluoroborate

97%

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

form

solid

reaction suitability

core: iron
reagent type: catalyst

mp

177.1-178.3 °C

storage temp.

−20°C

SMILES string

[Fe].F[B-](F)(F)F.[CH]1[CH][CH][CH][CH]1.CC(C)(C)[PH+]([C]2[CH][CH][CH][CH]2)C(C)(C)C

InChI

1S/C13H22P.C5H5.BF4.Fe/c1-12(2,3)14(13(4,5)6)11-9-7-8-10-11;1-2-4-5-3-1;2-1(3,4)5;/h7-10H,1-6H3;1-5H;;/q;;-1;/p+1

InChI key

WQLVZQWLDHMEBW-UHFFFAOYSA-O

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service