Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733644

Sigma-Aldrich

Poly(allylamine hydrochloride)

Mw 450,000

Synonym(s):

PAA HCl, PAH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH2NH2 · HCl)]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

Mw 450,000

transition temp

Tm 342-347 °C

storage temp.

2-8°C

SMILES string

Cl.NCC=C

InChI

1S/C3H7N/c1-2-3-4/h2H,1,3-4H2

InChI key

VVJKKWFAADXIJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to make redox hydrogel-modified electrodes for measuring enzyme responses.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiyao Liu et al.
Expert opinion on drug delivery, 9(10), 1197-1207 (2012-09-27)
Technologies to increase tissue vascularity are critically important to the fields of tissue engineering and cardiovascular medicine. Angiogenic factors, like VEGF, have been widely investigated to induce vascular endothelial cell proliferation and angiogenesis for establishing a vascular network. However, effective
L Minati et al.
International journal of pharmaceutics, 438(1-2), 45-52 (2012-09-11)
Gold nanoparticles functionalized with doxorubicin and stabilized with multilayers of degradable polyelectrolyte were allowed to age in aqueous medium in vitro in order to show the possibility of drug release in cellular environment. The chemico-physical characteristics of the nanoparticles are
Patrícia Pereira et al.
Journal of colloid and interface science, 387(1), 84-94 (2012-09-18)
The purpose of the present study is to provide a tool for an efficient design and synthesis of non-viral vectors for small RNA delivery. The effects of properties of the polycation, such as molecular weight, charge density and backbone structure
Xin Gao et al.
International journal of nanomedicine, 7, 4037-4051 (2012-08-14)
We designed dual-functional nanoparticles for in vivo application using a modified electrostatic and covalent layer-by-layer assembly strategy to address the challenge of assessment and treatment of hormone-refractory prostate cancer. Core-shell nanoparticles were formulated by integrating three distinct functional components, ie
Ilaria E Palamà et al.
Biotechnology and bioengineering, 110(2), 586-596 (2012-08-14)
Alignment of skeletal myoblasts is considered a critical step during myotube formation. The C2C12 cell line is frequently used as a model of skeletal muscle differentiation that can be induced by lowering the serum concentration in standard culture flasks. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service