Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733245

Sigma-Aldrich

Copper(II) acetate 1,2-bis(diphenylphosphino)ethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H30CuO4P2
CAS Number:
Molecular Weight:
580.05
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: copper
reagent type: catalyst

mp

120-125 °C

SMILES string

CC(=O)O[Cu]OC(C)=O.C(CP(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C26H24P2.2C2H4O2.Cu/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26;2*1-2(3)4;/h1-20H,21-22H2;2*1H3,(H,3,4);/q;;;+2/p-2

InChI key

RPIBQESUBINVPB-UHFFFAOYSA-L

General description

May contain up to 5% Tetrahydrofuran

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service