Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733008

Sigma-Aldrich

(S)-HuaCat

95%

Synonym(s):

(S)-N-(4-Dodecylphenylsulfonyl)pyrrolidine-2-carboxamide, (S)-N-(p-Dodecylphenylsulfonyl)-2-pyrrolidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H38N2O3S
CAS Number:
Molecular Weight:
422.62
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

optical activity

[α]/D −110.2°, c = 0.5 in chloroform

mp

203-208 °C

SMILES string

O=C(NS(=O)(C1=CC=C(CCCCCCCCCCCC)C=C1)=O)[C@H]2NCCC2

InChI

1S/C23H38N2O3S/c1-2-3-4-5-6-7-8-9-10-11-13-20-15-17-21(18-16-20)29(27,28)25-23(26)22-14-12-19-24-22/h15-18,22,24H,2-14,19H2,1H3,(H,25,26)/t22-/m0/s1

InChI key

FAGYODAJNLXMRF-QFIPXVFZSA-N

General description

Mixture of branched isomers on the dodecyl chain

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service