Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

732982

Sigma-Aldrich

7-Fluoro-4-hydroxycoumarin

Synonym(s):

7-Fluoro-4-hydroxy-1(2H)-benzopyran-2-one, 7-Fluoro-4-hydroxy-2H-chromen-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5FO3
CAS Number:
Molecular Weight:
180.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

220-225 °C

SMILES string

OC1=CC(=O)Oc2cc(F)ccc12

InChI

1S/C9H5FO3/c10-5-1-2-6-7(11)4-9(12)13-8(6)3-5/h1-4,11H

Inchi Key

FSWKHPJLMMGTBV-UHFFFAOYSA-N

Application

Reactant for:
  • Enantioselective synthesis of polycyclic coumarins via in situ formed primary amine-imine-catalyzed asymmetric Michael addition to cyclic enones
  • Preparation of dicoumarol analogs as inhibitors of human NAD(P)H quinone oxidoreductase-1 (NQO1) as agents active against human pancreatic and colon cancer cells
  • Imidazolidinecarboxylic acid-catalyzed asymmetric Michael reaction with unsaturated ketones in a highly atom-economic catalytic one-step formation of optically active warfarin anticoagulant
  • Asymmetric synthesis of functionalized α-keto esters and their hemiketal and hemiaminal derivatives via direct Michael addition with unsaturated α-keto esters catalyzed by bisoxazoline-copper(II) complexes

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service