Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

732664

Sigma-Aldrich

Methyl 1,3-dimethyl-1H-pyrazole-5-carboxylate

Synonym(s):

1,3-Dimethyl-1H-pyrazole-5-carboxylic acid methyl ester, Methyl 2,5-dimethyl-2H-pyrazole-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O2
CAS Number:
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

liquid

refractive index

n20/D 1.491

density

1.150 g/mL at 25 °C

SMILES string

COC(=O)c1cc(C)nn1C

InChI

1S/C7H10N2O2/c1-5-4-6(7(10)11-3)9(2)8-5/h4H,1-3H3

InChI key

KYQGPXFQWUSRMY-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids not in Storage Class 3

wgk_germany

WGK 1

flash_point_f

173.1 °F

flash_point_c

78.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service