Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

732354

Sigma-Aldrich

p-Toluic acid-(phenyl-13C6)

99 atom % 13C, 98% (CP)

Synonym(s):

4-Methylbenzoic acid-(phenyl-13C6)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313C6H4CO2H
Molecular Weight:
142.10
UNSPSC Code:
12352106

isotopic purity

99 atom % 13C

assay

98% (CP)

bp

274-275 °C (lit.)

mp

177-180 °C (lit.)

mass shift

M+6

SMILES string

C[13C]1=[13CH][13CH]=[13C](C(O)=O)[13CH]=[13CH]1

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service