Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

732206

Sigma-Aldrich

(1S,2S)-N-p-Tosyl-1,2-di(1-naphthyl)ethylenediamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H26N2O2S
CAS Number:
Molecular Weight:
466.59
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

97%

form

solid

mp

181-186 °C

functional group

amine

SMILES string

Cc1ccc(cc1)S(=O)(=O)N[C@H]([C@@H](N)c2cccc3ccccc23)c4cccc5ccccc45

InChI

1S/C29H26N2O2S/c1-20-16-18-23(19-17-20)34(32,33)31-29(27-15-7-11-22-9-3-5-13-25(22)27)28(30)26-14-6-10-21-8-2-4-12-24(21)26/h2-19,28-29,31H,30H2,1H3/t28-,29-/m0/s1

InChI key

GYVYYRQMNGSCRR-VMPREFPWSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Chin group is interested in computational and experimental approaches to understanding stereoselective recognition and catalysis. Their studies in weak forces (H-bonding, electronic and steric effects) has led to a highly efficient method for making limitless varieties of chiral vicinal diamines from the 'mother diamine' that are useful for developing stereoselective organocatalysts or transition metal-based catalysts as well as for developing drugs (Acc Chem Res (2012) p1345). The 'mother diamine' is also useful for making binol, monophos and binap analogs. The Chin group is also interested in using reversible covalent bonds for stereoselective recognition and L to D conversion of natural and non-natural amino acids (EJOC (2012) p229).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service