Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

732184

Sigma-Aldrich

Oxo[hexa(trifluoroacetato)]tetrazinc

contains ≤1 molar equivalent trifluoroacetic acid

Synonym(s):

ZnTAC24®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6F18O13Zn4
Molecular Weight:
961.70
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

Quality Level

contains

≤1 molar equivalent trifluoroacetic acid

reaction suitability

reagent type: catalyst
core: zinc

mp

>300 °C

SMILES string

OC(=O)C(F)(F)F.FC(F)(F)C1O[Zn]2OC(O[Zn]34OC(O[Zn](O1)OC(O[Zn](OC(O2)C(F)(F)F)(OC(O3)C(F)(F)F)O4)C(F)(F)F)C(F)(F)F)C(F)(F)F

InChI

1S/7C2HF3O2.O.4Zn/c7*3-2(4,5)1(6)7;;;;;/h(H,6,7);6*1H;;;;;/q;6*-2;;4*+3

InChI key

NOYQMFPDHODIRG-UHFFFAOYSA-N

Legal Information

Sold in collaboration with Takasago for research purposes only.
ZnTAC24 is a registered trademark of Takasago Intl. Corp.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service