Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731862

Sigma-Aldrich

CdSe/ZnS quantum dots

fluorescence λem 485 nm, 5 mg/mL in toluene/aliphatic amine

Synonym(s):

NanoBlue CdSe/ZnS - 485

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
26111700

form

liquid

Quality Level

availability

available only in USA

concentration

5 mg/mL in toluene/aliphatic amine

color

blue

density

0.866 g/mL at 20 °C (lit.)

fluorescence

FWHM <60 nm, quantum yield >80%
λem 485 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Nanosys, Inc. For research and development only; excluding use for biological assay systems and/or combinatorial chemistry.
NanoBlue is a trademark of Nanosys, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

48.2 °F

flash_point_c

9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service