Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731579

Sigma-Aldrich

2-(3-Oxetanylidene)acetonitrile

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO
CAS Number:
Molecular Weight:
95.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

form

solid

availability

not available in EU

mp

61-65 °C

storage temp.

2-8°C

SMILES string

N#C\C=C1\COC1

InChI

1S/C5H5NO/c6-2-1-5-3-7-4-5/h1H,3-4H2

InChI key

DSKHBLMKLTZNRU-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service