Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731455

Sigma-Aldrich

(3R,4E)-Methyl 3-(dimethylphenylsilyl)-4-hexenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O2Si
CAS Number:
Molecular Weight:
262.42
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

core: silicon
reagent type: catalyst

refractive index

n20/D 1.507

density

0.985 g/mL at 25 °C

functional group

ether

SMILES string

COC(=O)C[C@H](\C=C\C)[Si](C)(C)c1ccccc1

InChI

1S/C15H22O2Si/c1-5-9-14(12-15(16)17-2)18(3,4)13-10-7-6-8-11-13/h5-11,14H,12H2,1-4H3/b9-5+/t14-/m0/s1

InChI key

LCBVSVWXXPCVKB-KVEVGEHQSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service