Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

730726

Sigma-Aldrich

Triethylgallium

packaged for use in deposition systems

Synonym(s):

Et3Ga, TEGa, triethyl gallium, TEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)3Ga
CAS Number:
Molecular Weight:
156.91
EC Number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

liquid

reaction suitability

core: gallium

bp

109 °C/300 mmHg (lit.)

mp

-82 °C (lit.)

density

1.067 g/mL at 25 °C

InChI

1S/3C2H5.Ga/c3*1-2;/h3*1H2,2H3;

InChI key

RGGPNXQUMRMPRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 31 Gallium

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vossen, J. L.; Kern, W.
Thin Film Processes II, 1991, 866-866 null
M. Yamaguchi
Sci. Synth., 7, 387-387 (2004)
Nikolay Kornienko et al.
ACS nano, 9(4), 3951-3960 (2015-04-04)
The tunable physical and electronic structure of III-V semiconductor alloys renders them uniquely useful for a variety of applications, including biological imaging, transistors, and solar energy conversion. However, their fabrication typically requires complex gas phase instrumentation or growth from high-temperature

Articles

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

See All

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service