Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

730297

Sigma-Aldrich

TEMPO methacrylate

97%

Synonym(s):

4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl, PTMA monomer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22NO3
CAS Number:
Molecular Weight:
240.32
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

powder

mp

86-90 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OC1CC(C)(C)N([O])C(C)(C)C1

InChI

1S/C13H22NO3/c1-9(2)11(15)17-10-7-12(3,4)14(16)13(5,6)8-10/h10H,1,7-8H2,2-6H3

InChI key

BTWSPOZXDCFMLX-UHFFFAOYSA-N

General description

TEMPO methacrylate (4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl) undergoes anionic dispersion polymerization to afford nanometer-sized polymer particles bearing 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO) radicals.[1]

Application

Stable nitroxide radical useful in controlling living radical polymerizations, with a methacrylate functionality for cross-linking or orthogonal polymerization.
TEMPO methacrylate (4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl) may be employed as counter radical for polymerization of styrene.[2]

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Controlled radical polymerization of styrene in the presence of a polymerizable nitroxide compound.
Li C, et al.
Macromolecules, 32(21), 7012-7014 (1999)
Nakahara, K.; Iwasa, S.; Satoh, M.; Morioka, Y.; Iriyama, J.; Suguro, M.; Hasegawa, E.
Chemical Physics Letters, 359, 351-351 (2002)
Magnetic force microscopic images of nanometer-sized polyradical particles.
Michinobu T, et al.
Polymer Journal, 35(1), 71-75 (2003)
Battery-inspired, nonvolatile, and rewritable memory architecture: a radical polymer-based organic device.
Yasunori Yonekuta et al.
Journal of the American Chemical Society, 129(46), 14128-14129 (2007-10-30)

Articles

Block copolymer synthesis using a commercially available nitroxide-mediated radical polymerization (NMP) initiator

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service