Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

730262

Sigma-Aldrich

1,4-Bis(4-vinylphenoxy)butane

>90%

Synonym(s):

JandaJel crosslinker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O2
CAS Number:
Molecular Weight:
294.39
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

>90%

form

solid

mp

123-128 °C
130-133 °C (lit.)

storage temp.

2-8°C

SMILES string

C=Cc1ccc(OCCCCOc2ccc(C=C)cc2)cc1

InChI

1S/C20H22O2/c1-3-17-7-11-19(12-8-17)21-15-5-6-16-22-20-13-9-18(4-2)10-14-20/h3-4,7-14H,1-2,5-6,15-16H2

InChI key

RAPXVKLHABTPOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

JandaJel is a trademark of Sigma-Aldrich Co. LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service