729175
Lanthanum nickelate
nanopowder, <100 nm particle size (BET)
Synonym(s):
LaNO, Lanthanum nickel oxide
About This Item
Recommended Products
form
nanopowder
composition
La2NiO4
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
surface area
5-10 m2/g
particle size
<100 nm (BET)
mp
>400 °C
application(s)
battery manufacturing
greener alternative category
SMILES string
O=[La]O[Ni]O[La]=O
InChI
1S/2La.Ni.4O
InChI key
VJMGRMCCSVDPHO-UHFFFAOYSA-N
General description
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service