Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

728578

Sigma-Aldrich

(RS)-1-Methyl-2-propynyl p-toluenesulfonate

95%

Synonym(s):

1-Butyn-3-yl tosylate, 3-Butyn-2-ol 4-methylbenzene sulfonate, Toluene-4-sulfonic acid 1-methyl-2-propynyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O3S
CAS Number:
Molecular Weight:
224.28
Beilstein/REAXYS Number:
2650656
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥95.0% (HPLC)
95%

form

powder

Storage temp.

2-8°C

SMILES string

CC(OS(=O)(=O)c1ccc(C)cc1)C#C

InChI

1S/C11H12O3S/c1-4-10(3)14-15(12,13)11-7-5-9(2)6-8-11/h1,5-8,10H,2-3H3

Inchi Key

XWRDBUBKBUYLQI-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the Stereochemistry of the Aromatic Claisen Rearrangement. Thermal Rearrangement of erythroid and threoid ortho-Dienones.
Wunderli, A. et al.,
Helvetica Chimica Acta, 60, 2436-2459 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service