Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

728195

Sigma-Aldrich

1,4-Diphenyl-2-butyne-1,4-dione

≥95.0% (HPLC)

Synonym(s):

Dibenzoylacetylene, Dibenzoylethyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10O2
CAS Number:
Molecular Weight:
234.25
Beilstein/REAXYS Number:
611463
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥95.0% (HPLC)

form

crystals

storage temp.

2-8°C

SMILES string

O=C(C#CC(=O)c1ccccc1)c2ccccc2

InChI

1S/C16H10O2/c17-15(13-7-3-1-4-8-13)11-12-16(18)14-9-5-2-6-10-14/h1-10H

InChI key

BLCJVQFFSUNDMV-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service