Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

728179

Sigma-Aldrich

3-(1-Piperazinyl)propionic acid

98%

Synonym(s):

3-Piperazinopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O2
CAS Number:
Molecular Weight:
158.20
Beilstein/REAXYS Number:
125453
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (HPLC)
98%

form

crystals

reaction suitability

reaction type: solution phase peptide synthesis

SMILES string

OC(=O)CCN1CCNCC1

InChI

1S/C7H14N2O2/c10-7(11)1-4-9-5-2-8-3-6-9/h8H,1-6H2,(H,10,11)

InChI key

RSEHHYMPNZWTGQ-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service