Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727350

Sigma-Aldrich

Methyl (R)-3-aminobutyrate p-toluenesulfonate salt

ChiPros®, produced by BASF, ≥97.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO2 · C7H8O3S
CAS Number:
Molecular Weight:
289.35
Beilstein/REAXYS Number:
9874258
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

assay

≥97.5% (GC)

form

lumps

optical purity

optical purity: ≥97.5%

SMILES string

COC(=O)C[C@@H](C)N.Cc1ccc(cc1)S(O)(=O)=O

InChI

1S/C7H8O3S.C5H11NO2/c1-6-2-4-7(5-3-6)11(8,9)10;1-4(6)3-5(7)8-2/h2-5H,1H3,(H,8,9,10);4H,3,6H2,1-2H3/t;4-/m.1/s1

InChI key

BGTUHDOWLHXQIA-FZSMXKCYSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>212.0 °F

flash_point_c

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service