Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727245

Sigma-Aldrich

(S)-1-Cyclopropylethylamine

ChiPros®, produced by BASF, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

functional group

amine

SMILES string

C[C@H](N)C1CC1

InChI

1S/C5H11N/c1-4(6)5-2-3-5/h4-5H,2-3,6H2,1H3/t4-/m0/s1

InChI key

IXCXVGWKYIDNOS-BYPYZUCNSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

<69.8 °F

flash_point_c

< 21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service