Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727180

Sigma-Aldrich

(R)-2-Aminononane

produced by BASF, ChiPros®

Synonym(s):

(R)-2-Nonylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21N
CAS Number:
Molecular Weight:
143.27
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

≥98.5%

form

liquid

optical purity

ee: ≥98.0%

functional group

amine

SMILES string

CCCCCCC[C@@H](C)N

InChI

1S/C9H21N/c1-3-4-5-6-7-8-9(2)10/h9H,3-8,10H2,1-2H3/t9-/m1/s1

InChI key

ALXIFCUEJWCQQL-SECBINFHSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

165.2 °F

flash_point_c

74 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service