Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727121

Sigma-Aldrich

(R)-2-Methoxy-α-methylbenzylamine

ChiPros®, produced by BASF, ≥99.0%

Synonym(s):

(R)-1-(2-Methoxyphenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
Beilstein/REAXYS Number:
4840675
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

assay

≥99.0% (GC)
≥99.0%

form

liquid

optical purity

enantiomeric excess: ≥99.0%

SMILES string

COc1ccccc1[C@@H](C)N

InChI

1S/C9H13NO/c1-7(10)8-5-3-4-6-9(8)11-2/h3-7H,10H2,1-2H3/t7-/m1/s1

InChI key

VENQOHAPVLVQKV-SSDOTTSWSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Skin Corr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service