Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726958

Sigma-Aldrich

(S)-3-Bromo-α-methylbenzylamine

Chipros®, produced by BASF, ≥99.0%

Synonym(s):

(S)-1-(3-Bromophenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BrN
CAS Number:
Molecular Weight:
200.08
Beilstein/REAXYS Number:
4798483
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

assay

≥99.0% (GC)
≥99.0%

form

liquid

optical purity

enantiomeric excess: ≥99.0%

SMILES string

C[C@H](N)c1cccc(Br)c1

InChI

1S/C8H10BrN/c1-6(10)7-3-2-4-8(9)5-7/h2-6H,10H2,1H3/t6-/m0/s1

InChI key

LIBZHYLTOAGURM-LURJTMIESA-N

Legal Information

ChiPros is a registered trademark of BASF SE

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service