Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726818

Sigma-Aldrich

(S)-(−)-N-Benzyl-α-methylbenzylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-(−)-N-(1-Phenylethyl)benzylamine, (S)-(−)-N-Benzyl-α-phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NHCH2C6H5
CAS Number:
Molecular Weight:
211.30
Beilstein/REAXYS Number:
3650264
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

refractive index

n20/D 1.563 (lit.)

bp

171 °C/15 mmHg (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

C[C@H](NCc1ccccc1)c2ccccc2

InChI

1S/C15H17N/c1-13(15-10-6-3-7-11-15)16-12-14-8-4-2-5-9-14/h2-11,13,16H,12H2,1H3/t13-/m0/s1

InChI key

ZYZHMSJNPCYUTB-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service