Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726699

Sigma-Aldrich

(R)-(+)-(3-Chlorophenyl)oxirane

ChiPros®, produced by BASF, 98%

Synonym(s):

(R)-(+)-3-Chlorostyrene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO
CAS Number:
Molecular Weight:
154.59
Beilstein/REAXYS Number:
3588590
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥97.5% (GC)
98%

form

liquid

optical purity

enantiomeric excess: ≥97.5%

density

1.217 g/mL at 20 °C (lit.)

SMILES string

Clc1cccc(c1)[C@@H]2CO2

InChI

1S/C8H7ClO/c9-7-3-1-2-6(4-7)8-5-10-8/h1-4,8H,5H2/t8-/m0/s1

InChI key

YVMKRPGFBQGEBF-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

206.6 °F - closed cup

flash_point_c

97 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service