Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726567

Sigma-Aldrich

(S)-(+)-2-Pentanol

ChiPros®, produced by BASF, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
88.15
Beilstein/REAXYS Number:
1718820
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

vapor density

3 (vs air)

assay

≥97.5% (GC)
98%

form

liquid

optical purity

enantiomeric excess: ≥97.5%

autoignition temp.

657 °F

expl. lim.

9 %

refractive index

n20/D 1.406 (lit.)

bp

118-119 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

SMILES string

CCC[C@H](C)O

InChI

1S/C5H12O/c1-3-4-5(2)6/h5-6H,3-4H2,1-2H3/t5-/m0/s1

InChI key

JYVLIDXNZAXMDK-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

93.2 °F - closed cup

flash_point_c

34 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service