Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

726095

Sigma-Aldrich

(±)-Diethyl trans-4-cyclohexene-1,2-dicarboxylate

greener alternative

≥98.0%

Synonym(s):

trans-4-Cyclohexene-1,2-dicarboxylic acid diethyl ester, Diethyl trans-1,2,3,6-tetrahydrophthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O4
CAS Number:
Molecular Weight:
226.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (GC)
≥98.0%

form

liquid

greener alternative product score

old score: 8
new score: 3
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

CCOC(=O)[C@@H]1CC=CC[C@H]1C(=O)OCC

InChI

1S/C12H18O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-6,9-10H,3-4,7-8H2,1-2H3/t9-,10-/m1/s1

InChI key

OWVOHDKOANTMJU-NXEZZACHSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy”, “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstocks”. Click here to view its DOZN scorecard.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Burgos, G.J. Ellames
Journal of Labelled Compounds & Radiopharmaceuticals, XLI, 443-449 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service