Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

725935

Sigma-Aldrich

Fmoc-β-Lys(Boc)-OH

≥98% (HPLC)

Synonym(s):

(S)-6-(tert-Butoxycarbonylamino)-3-(fluoren-9-ylmethoxycarbonylamino)hexanoic acid, Nβ-Fmoc-Nϖ-Boc-L-β-lysine, Fmoc-L-β-HomoOrn(Boc)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N2O6
CAS Number:
Molecular Weight:
468.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -8.5±1.0°, c = 1 in chloroform

composition

carbon, 65.9-67.5%
nitrogen, 5.6-6.4%

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCC[C@@H](CC(O)=O)NC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C26H32N2O6/c1-26(2,3)34-24(31)27-14-8-9-17(15-23(29)30)28-25(32)33-16-22-20-12-6-4-10-18(20)19-11-5-7-13-21(19)22/h4-7,10-13,17,22H,8-9,14-16H2,1-3H3,(H,27,31)(H,28,32)(H,29,30)/t17-/m0/s1

InChI key

SETTXXCZEDLMRX-KRWDZBQOSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 4 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service