725803
Ethyl 4,4-difluorovalerate
≥95.0%
Synonym(s):
Ethyl 4,4-difluoropentanoate
About This Item
Recommended Products
assay
≥95.0%
form
liquid
SMILES string
CCOC(=O)CCC(C)(F)F
InChI
1S/C7H12F2O2/c1-3-11-6(10)4-5-7(2,8)9/h3-5H2,1-2H3
InChI key
BMPDDDGLNXKZFD-UHFFFAOYSA-N
Packaging
signalword
Danger
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
supp_hazards
Storage Class
3 - Flammable Liquids
wgk_germany
WGK 3
flash_point_f
131.0 °F
flash_point_c
55 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service