Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

725579

Sigma-Aldrich

[11-(Methylcarbonylthio)undecyl]tri(ethylene glycol) acetic acid

95%

Synonym(s):

[1-(Methylcarbonylthio)undec-11-yl]tri(ethylene glycol) acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H40O7S
CAS Number:
Molecular Weight:
436.60
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

95%

form

solid

mp

28-32 °C

storage temp.

2-8°C

SMILES string

OC(=O)COCCOCCOCCOCCCCCCCCCCCSC=O

InChI

1S/C20H38O7S/c21-19-28-17-9-7-5-3-1-2-4-6-8-10-24-11-12-25-13-14-26-15-16-27-18-20(22)23/h19H,1-18H2,(H,22,23)

InChI key

HQIKYCFDGNDPNW-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Protocols

Hydrolyzing agents deprotect protected thiol derivatives, obtaining unstable free thiols during complex compound synthesis for self-assembled monolayers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service