Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

725552

Sigma-Aldrich

[11-(Methylcarbonylthio)undecyl]tri(ethylene glycol) methyl ether

96%

Synonym(s):

[1-(Methylcarbonylthio)undec-11-yl]tri(ethylene glycol)monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40O5S
Molecular Weight:
392.59
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

96%

form

liquid

refractive index

n20/D 1.469

density

0.976 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

COCCOCCOCCOCCCCCCCCCCCSC(C)=O

InChI

1S/C20H40O5S/c1-20(21)26-19-11-9-7-5-3-4-6-8-10-12-23-15-16-25-18-17-24-14-13-22-2/h3-19H2,1-2H3

InChI key

JXIPMJZDYPBSOO-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service