Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

724416

Sigma-Aldrich

1,2-Dimethyl-3-propylimidazolium bis(trifluoromethylsulfonyl)imide

for energy applications

Synonym(s):

DMPIIm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15F6N3O4S2
CAS Number:
Molecular Weight:
419.36
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

assay

99.9% trace metals basis

form

liquid

impurities

≤0.5% H2O

refractive index

n20/D 1.433

mp

15 °C (lit.)

density

1.436 g/mL at 25 °C

application(s)

battery manufacturing

SMILES string

CCC[n+]1ccn(C)c1C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C8H15N2.C2F6NO4S2/c1-4-5-10-7-6-9(3)8(10)2;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h6-7H,4-5H2,1-3H3;/q+1;-1

InChI key

XOZHIVUWCICHSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Navarra, M. A.; Panero, S.; Scrosati, B.
Electrochemical and Solid-State Letters, 8, A324-A324 (2005)
Koch, V. R.; Nanjundiah, C.; Appetecchi, G. Battista; Scrosati, B.
Journal of the Electrochemical Society, 142, L116-L116 (1995)
Shiro Seki et al.
The journal of physical chemistry. B, 110(21), 10228-10230 (2006-05-26)
Highly reversible, safe lithium secondary batteries that use imidazolium-cation-based room-temperature ionic liquid as an electrolyte and lithium metal as an anode material were realized by the molecular design. To achieve higher reduction stability, an electron-donating substituent was introduced to promote
Ueki, Takeshi; Arai, Asako Ayusawa; Kodama, Kiochi
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 81, 1829-1829 (2009)

Articles

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Lithium-ion batteries offer high energy density and cyclic performance for portable electronic devices.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service