Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

724386

1-Bromopentane-13C5

≥99 atom % 13C, ≥99% (CP)

Synonym(s):

1-Pentyl bromide-13C5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C5H11Br
Molecular Weight:
156.01
UNSPSC Code:
12352101
Pricing and availability is not currently available.

isotopic purity

≥99 atom % 13C

assay

≥99% (CP)

mass shift

M+5

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

89.6 °F - closed cup

flash_point_c

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service