Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

722553

Sigma-Aldrich

Tos-Pro-OH

97%

Synonym(s):

(S)-N-Toluenesulfonylpyrrolidine-2-carboxylic acid, N-p-Tosyl-L-proline, N-Toluenesulfonyl-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO4S
CAS Number:
Molecular Weight:
269.32
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

80-90 °C

application(s)

peptide synthesis

SMILES string

Cc1ccc(cc1)S(=O)(=O)N2CCC[C@H]2C(O)=O

InChI

1S/C12H15NO4S/c1-9-4-6-10(7-5-9)18(16,17)13-8-2-3-11(13)12(14)15/h4-7,11H,2-3,8H2,1H3,(H,14,15)/t11-/m0/s1

InChI key

CGPHGPCHVUSFFA-NSHDSACASA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service