Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

721514

Sigma-Aldrich

(S)-1-Boc-3-fluoropyrrolidine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16FNO2
CAS Number:
Molecular Weight:
189.23
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.441

density

1.069 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)N1CC[C@H](F)C1

InChI

1S/C9H16FNO2/c1-9(2,3)13-8(12)11-5-4-7(10)6-11/h7H,4-6H2,1-3H3/t7-/m0/s1

InChI key

SUECTKVSIDXQQE-ZETCQYMHSA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

217.9 °F

flash_point_c

103.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service