Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720933

Sigma-Aldrich

Potassium 3-methyl-2-buten-2-yltrifluoroborate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9BF3K
Molecular Weight:
176.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

218-223 °C

storage temp.

2-8°C

SMILES string

[K+].C\C(C)=C(\C)[B-](F)(F)F

InChI

1S/C5H9BF3.K/c1-4(2)5(3)6(7,8)9;/h1-3H3;/q-1;+1

InChI key

GYUUSPIOJSPUJM-UHFFFAOYSA-N

Application

Potassium 3-methyl-2-buten-2-yltrifluoroborate can be used to prepare ethyl 2-[(4-bromophenyl)amino]-3,4-dimethylpent-3-enoate by reacting with (E)-ethyl 2-(4-bromophenylimino)acetate and ytterbium triflate.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julio Zukerman-Schpector et al.
Acta crystallographica. Section E, Structure reports online, 70(Pt 10), o1122-o1123 (2014-12-09)
In the title compound, C15H20BrNO2, there are two independent mol-ecules (A and B) comprising the asymmetric unit and these adopt very similar conformations. In A, the dihedral angle between the CO2 and MeC=CMe2 groups is 80.7 (3)°, and these make dihedral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service