Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720860

Sigma-Aldrich

4-Bromo-1-methylindole

95%

Synonym(s):

4-Bromo-1-methyl-1H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8BrN
CAS Number:
Molecular Weight:
210.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.636

density

1.456 g/mL at 25 °C

functional group

bromo

SMILES string

Cn1ccc2c(Br)cccc12

InChI

1S/C9H8BrN/c1-11-6-5-7-8(10)3-2-4-9(7)11/h2-6H,1H3

InChI key

JZOSXTYDJPHXQD-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service