720194
7-Bromo-6-chloro-7-deazapurine
95%
Synonym(s):
5-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H3BrClN3
CAS Number:
Molecular Weight:
232.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
assay
95%
form
solid
mp
221-225 °C
SMILES string
Clc1ncnc2[nH]cc(Br)c12
InChI
1S/C6H3BrClN3/c7-3-1-9-6-4(3)5(8)10-2-11-6/h1-2H,(H,9,10,11)
InChI key
OXLMTRZWMHIZBY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
pcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service