Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720151

Sigma-Aldrich

(S)-2-(2-Bromophenyl)-4-isopropyl-5,5-dimethyl-4,5-dihydrooxazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18BrNO
Molecular Weight:
296.20
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

liquid

optical activity

[α]22/D −41.0°, c = 0.5 in chloroform

refractive index

n20/D 1.541

density

1.247 g/mL at 25 °C

functional group

bromo
ether

SMILES string

CC1(C)[C@H](C(C)C)N=C(C2=CC=CC=C2Br)O1

InChI

1S/C14H18BrNO/c1-9(2)12-14(3,4)17-13(16-12)10-7-5-6-8-11(10)15/h5-9,12H,1-4H3/t12-/m0/s1

InChI key

SJKCCGPXYWNYGX-LBPRGKRZSA-N

Application

(S)-2-(2-Bromophenyl)-4-isopropyl-5,5-dimethyl-4,5-dihydrooxazole can be used as a key intermediate for the preparation of (S)-5,5-(dimethyl)-i-Pr-PHOX, a chiral ligand employed in the palladium-catalyzed asymmetric transformations.[1]

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, Synthesis, and Applications of Potential Substitutes of t-Bu-Phosphinooxazoline in Pd-Catalyzed Asymmetric Transformations and Their Use for the Improvement of the Enantioselectivity in the Pd-Catalyzed Allylation Reaction of Fluorinated Allyl Enol Carbonates
Belanger E, et al.
The Journal of Organic Chemistry, 77(1), 317-331 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service