Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720135

Sigma-Aldrich

4-Bromo-1-(2-tetrahydropyranyl)pyrazole

96%

Synonym(s):

4-Bromo-(N-tetrahydropyranyl)pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11BrN2O
CAS Number:
Molecular Weight:
231.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

density

1.493 g/mL at 25 °C

SMILES string

Brc1cnn(c1)C2CCCCO2

InChI

1S/C8H11BrN2O/c9-7-5-10-11(6-7)8-3-1-2-4-12-8/h5-6,8H,1-4H2

InChI key

HDNCEUNJWJHWMO-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service